Product Details

SNP ID
rs148756611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113422916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGGTACAAGCCTTTGGGAACG[A/C]GGTCCAGGCCGTACAGCAACCCGAA
Phenotype
MIM: 610620
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADPRHL1 PubMed Links

Gene Details

Gene
ADPRHL1
Gene Name
ADP-ribosylhydrolase like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138430.4 1058 Missense Mutation CGC,CTC R324L NP_612439.2
NM_199162.2 1058 Missense Mutation CGC,CTC R242L NP_954631.1
Gene
LOC101928841
Gene Name
collagen alpha-1(II) chain-like
There are no transcripts associated with this gene.

View Full Product Details