Product Details

SNP ID
rs148914536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:23330502 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGAAGGTCATTCCTGGCAGCTGA[A/G]AAGTTTGCTCTGGCTTGTCTTAGCC
Phenotype
MIM: 604490 MIM: 608896
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SACS PubMed Links

Gene Details

Gene
SACS
Gene Name
sacsin molecular chaperone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278055.1 13458 Silent Mutation NP_001264984.1
NM_014363.5 13458 Silent Mutation NP_055178.3
XM_005266338.2 13458 Silent Mutation XP_005266395.1
XM_011535039.2 13458 Silent Mutation XP_011533341.1
XM_017020539.1 13458 Missense Mutation XP_016876028.1
Gene
SGCG
Gene Name
sarcoglycan gamma
There are no transcripts associated with this gene.

View Full Product Details