Product Details

SNP ID
rs149372773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:95433941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAGTGACCACGCGAGCCTCCTC[A/G]GTGATAACAGCCACTTGGGTTTACC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLDN10 PubMed Links

Gene Details

Gene
CLDN10
Gene Name
claudin 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160100.1 343 Silent Mutation TCA,TCG S36S NP_001153572.1
NM_006984.4 343 Intron NP_008915.1
NM_182848.3 343 Silent Mutation TCA,TCG S36S NP_878268.1
XM_011521134.2 343 Intron XP_011519436.1
XM_017020843.1 343 Silent Mutation TCA,TCG S36S XP_016876332.1
XM_017020844.1 343 Silent Mutation TCA,TCG S36S XP_016876333.1

View Full Product Details