Product Details

SNP ID
rs149877405
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:47943398 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTGGCAACTGAAATTTCACATC[C/T]ACATGTGCTTGCTTCGCTAAGGTCA
Phenotype
MIM: 603921
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SUCLA2 PubMed Links

Gene Details

Gene
SUCLA2
Gene Name
succinate-CoA ligase ADP-forming beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003850.2 1642 Silent Mutation GTA,GTG V455V NP_003841.1
XM_017020806.1 1642 Silent Mutation GTA,GTG V397V XP_016876295.1
XM_017020807.1 1642 Silent Mutation GTA,GTG V376V XP_016876296.1
XM_017020808.1 1642 Intron XP_016876297.1
XM_017020809.1 1642 Silent Mutation GTA,GTG V321V XP_016876298.1

View Full Product Details