Product Details

SNP ID
rs150015002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40933633 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTTGATAACTGAAGTGATTACA[A/G]TGCCAGGTGGGTGAGCAACCAGCTG
Phenotype
MIM: 189973
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ELF1 PubMed Links

Gene Details

Gene
ELF1
Gene Name
E74 like ETS transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145353.1 2254 Missense Mutation ACT,ATT T527I NP_001138825.1
NM_172373.3 2254 Missense Mutation ACT,ATT T551I NP_758961.1
XM_005266276.1 2254 Missense Mutation ACT,ATT T551I XP_005266333.1
XM_005266277.3 2254 Missense Mutation ACT,ATT T551I XP_005266334.1
XM_011534950.1 2254 Missense Mutation ACT,ATT T551I XP_011533252.1
XM_011534951.2 2254 Missense Mutation ACT,ATT T492I XP_011533253.1
XM_017020409.1 2254 Missense Mutation ACT,ATT T551I XP_016875898.1
XM_017020410.1 2254 Missense Mutation ACT,ATT T551I XP_016875899.1
XM_017020411.1 2254 Missense Mutation ACT,ATT T551I XP_016875900.1
XM_017020412.1 2254 Missense Mutation ACT,ATT T551I XP_016875901.1
XM_017020413.1 2254 Missense Mutation ACT,ATT T551I XP_016875902.1

View Full Product Details