Product Details

SNP ID
rs150664551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:83879531 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTGTCTGTAGGGAATTAATCTC[G/T]GACGCGGAGGAGTTGGCATCTCGTC
Phenotype
MIM: 609678
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLITRK1 PubMed Links

Gene Details

Gene
SLITRK1
Gene Name
SLIT and NTRK like family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281503.1 2863 Silent Mutation TCA,TCC S659S NP_001268432.1
NM_052910.2 2863 Silent Mutation TCA,TCC S659S NP_443142.1

View Full Product Details