Product Details

SNP ID
rs150680500
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:61410362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATATATAACCCAAGTATAAAGTA[C/T]GACCAACAAGTTGTGAAAGCACTAA
Phenotype
MIM: 614449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH20 PubMed Links

Gene Details

Gene
PCDH20
Gene Name
protocadherin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022843.3 5537 UTR 3 NP_073754.2
XM_017020708.1 5537 UTR 3 XP_016876197.1

View Full Product Details