Product Details

SNP ID
rs138037936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:38254899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAGAATTAAACTTGGAAATCACG[C/T]TCCCTGATGGGGTGATAGTGGCCTT
Phenotype
MIM: 616845
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLEC14A PubMed Links

Gene Details

Gene
CLEC14A
Gene Name
C-type lectin domain family 14 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175060.2 1472 Missense Mutation AAC,AGC N375S NP_778230.1

View Full Product Details