Product Details

SNP ID
rs138110777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24573749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTCTGGAGGAACCCCTGACGAC[C/T]TTCCATAGGAGACGATGCCGTGGGC
Phenotype
MIM: 116830
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTSG PubMed Links

Gene Details

Gene
CTSG
Gene Name
cathepsin G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001911.2 693 Missense Mutation AAG,AGG K219R NP_001902.1
XM_011536499.1 693 Missense Mutation AAG,AGG K233R XP_011534801.1

View Full Product Details