Product Details

SNP ID
rs138153264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90275466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCTTTTCCTCATTGGTAATAGC[C/T]GAGCGTTTTCCTCTAGAGGGACAGT
Phenotype
MIM: 602706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRDE2 PubMed Links

Gene Details

Gene
NRDE2
Gene Name
NRDE-2, necessary for RNA interference, domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017970.3 Intron NP_060440.2
XM_011536886.1 Intron XP_011535188.1
XM_017021394.1 Intron XP_016876883.1
XM_017021395.1 Intron XP_016876884.1
XM_017021396.1 Intron XP_016876885.1
Gene
PSMC1
Gene Name
proteasome 26S subunit, ATPase 1
There are no transcripts associated with this gene.

View Full Product Details