Product Details

SNP ID
rs138494163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20014514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGTGCTACCGGCAGCACGCTGT[C/G]TGATAGCGAGGAGGATCACGGTGTA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR4K14 PubMed Links

Gene Details

Gene
OR4K14
Gene Name
olfactory receptor family 4 subfamily K member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004712.1 680 Missense Mutation ACA,AGA T227R NP_001004712.1

View Full Product Details