Product Details

SNP ID
rs138513424
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100537308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTCAGGGGGCGGACCCGGGGGC[A/G]GAGATGAGCACCGGCCGCACTGGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BEGAIN PubMed Links

Gene Details

Gene
BEGAIN
Gene Name
brain enriched guanylate kinase associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159531.1 2731 UTR 3 NP_001153003.1
NM_020836.3 2731 UTR 3 NP_065887.1
XM_005267919.3 2731 UTR 3 XP_005267976.1
XM_005267920.4 2731 UTR 3 XP_005267977.1
XM_005267921.3 2731 UTR 3 XP_005267978.1
XM_011537031.1 2731 UTR 3 XP_011535333.1
XM_017021521.1 2731 UTR 3 XP_016877010.1
XM_017021522.1 2731 UTR 3 XP_016877011.1
Gene
WDR25
Gene Name
WD repeat domain 25
There are no transcripts associated with this gene.

View Full Product Details