Product Details

SNP ID
rs138605177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63312948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCCTCAGATGGTCTCACACTCCG[C/T]GGTGGCAGTGGAGCAGGCTCCGCAG
Phenotype
MIM: 609652
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPHB5 PubMed Links

Gene Details

Gene
GPHB5
Gene Name
glycoprotein hormone beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145171.3 430 Missense Mutation ACG,GCG T125A NP_660154.3

View Full Product Details