Product Details

SNP ID
rs138781112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105488478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCACCTTCTCTGCAGGCTTTGG[A/G]CGGGGCGGAGCCGAGAGCCACACTT
Phenotype
MIM: 123875 MIM: 601183
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf80 PubMed Links

Gene Details

Gene
C14orf80
Gene Name
chromosome 14 open reading frame 80
There are no transcripts associated with this gene.

Gene
CRIP1
Gene Name
cysteine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001311.4 315 Silent Mutation GGA,GGG G67G NP_001302.1
Gene
CRIP2
Gene Name
cysteine rich protein 2
There are no transcripts associated with this gene.

View Full Product Details