Product Details

SNP ID
rs138853366
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:80484950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAAAGGGAAAGGGTAGAGAAAT[C/T]TGACCTCAATTGTGGGCATTGGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEP128 PubMed Links

Gene Details

Gene
CEP128
Gene Name
centrosomal protein 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152446.3 Intron NP_689659.2
XM_006720056.3 Intron XP_006720119.1
XM_011536490.2 Intron XP_011534792.1
XM_011536491.2 Intron XP_011534793.1
XM_011536492.2 Intron XP_011534794.1
XM_011536493.2 Intron XP_011534795.1
XM_011536495.2 Intron XP_011534797.1
XM_017021042.1 Intron XP_016876531.1
XM_017021043.1 Intron XP_016876532.1

View Full Product Details