Product Details

SNP ID
rs139351024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20641044 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGGCCGTAGGACACAGCAGT[G/T]ATCAGCAAGGAAGATACAATGACGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR6S1 PubMed Links

Gene Details

Gene
OR6S1
Gene Name
olfactory receptor family 6 subfamily S member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001968.1 648 Silent Mutation NP_001001968.1

View Full Product Details