Product Details
- SNP ID
-
rs139700002
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24506158 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACCTCTTGCAGAGTGTCTGAGCCC[G/T]GCTTCAACACACCTGTTCTTCCCCA
- Phenotype
-
MIM: 118938
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CMA1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5249] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CMA1
- Gene Name
- chymase 1
- Gene
- LOC101927045
- Gene Name
- uncharacterized LOC101927045
There are no transcripts associated with this gene.
View Full Product Details