Product Details

SNP ID
rs139827649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20034076 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTTAGAGGATCGACTAGCAGCA[C/T]GGTACCTAACTGAGAATATTATGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4K13 PubMed Links

Gene Details

Gene
OR4K13
Gene Name
olfactory receptor family 4 subfamily K member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004714.1 683 Missense Mutation CAT,CGT H228R NP_001004714.1

View Full Product Details