Product Details

SNP ID
rs139896732
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:29577259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTCAGAGGATGCTGACACGCTC[A/C]CCGAGGGCTTTCATTTCTGTTTCTT
Phenotype
MIM: 605435
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRKD1 PubMed Links

Gene Details

Gene
PRKD1
Gene Name
protein kinase D1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002742.2 5279 Silent Mutation GGG,GGT G906G NP_002733.2
XM_005267859.1 5279 Silent Mutation GGG,GGT G914G XP_005267916.1
XM_011536964.2 5279 Silent Mutation GGG,GGT G838G XP_011535266.1
XM_011536965.2 5279 Silent Mutation GGG,GGT G818G XP_011535267.1
XM_017021461.1 5279 Silent Mutation GGG,GGT G818G XP_016876950.1
XM_017021462.1 5279 Silent Mutation GGG,GGT G741G XP_016876951.1

View Full Product Details