Product Details

SNP ID
rs140088005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60644760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTCACTAATTTTAAAAGCTAA[A/G]TACAGAAGATACATCATTTTGCCCC
Phenotype
MIM: 601205
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105378189 PubMed Links

Gene Details

Gene
LOC105378189
Gene Name
uncharacterized LOC105378189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011537451.2 2619 Intron XP_011535753.1
Gene
SALRNA1
Gene Name
senescence associated long non-coding RNA 1
There are no transcripts associated with this gene.

Gene
SIX1
Gene Name
SIX homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005982.3 2619 UTR 3 NP_005973.1
XM_017021602.1 2619 UTR 3 XP_016877091.1

View Full Product Details