Product Details

SNP ID
rs140355932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:94128611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATCTTGGCTGCTATGGAGGACGC[A/G]GCGATTCCTGCCCCAGTGAAGCCCA
Phenotype
MIM: 611319
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI27L2 PubMed Links

Gene Details

Gene
IFI27L2
Gene Name
interferon alpha inducible protein 27 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032036.2 158 Silent Mutation GCC,GCT A34A NP_114425.1

View Full Product Details