Product Details

SNP ID
rs140549878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49693278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACGGAAACTTCCTCTACGTGTG[A/G]GGGGGCTACGTGGTAAGGGGAAGAG
Phenotype
MIM: 611281 MIM: 602670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLHDC1 PubMed Links

Gene Details

Gene
KLHDC1
Gene Name
kelch domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172193.2 174 Missense Mutation TGA,TGG *28W NP_751943.1
XM_011536422.2 174 Nonsense Mutation TGA,TGG *28W XP_011534724.1
Gene
POLE2
Gene Name
DNA polymerase epsilon 2, accessory subunit
There are no transcripts associated with this gene.

View Full Product Details