Product Details

SNP ID
rs140558508
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100729017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCACGGACTCTGTGGAGAACC[C/T]GGGCAGTGCATTTGCACCGACGGCT
Phenotype
MIM: 176290
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLK1 PubMed Links

Gene Details

Gene
DLK1
Gene Name
delta like non-canonical Notch ligand 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317172.1 417 Silent Mutation CCC,CCT P71P NP_001304101.1
NM_003836.6 417 Silent Mutation CCC,CCT P71P NP_003827.3

View Full Product Details