Product Details

SNP ID
rs140610133
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21058256 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGTACCCAAACTGCAGGTACAAA[A/G]AGAAGCACCTGAACACACCTTACAT
Phenotype
MIM: 610018 MIM: 605272 MIM: 612485
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF40 PubMed Links

Gene Details

Gene
ARHGEF40
Gene Name
Rho guanine nucleotide exchange factor 40
There are no transcripts associated with this gene.

Gene
NDRG2
Gene Name
NDRG family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282211.1 435 Intron NP_001269140.1
NM_001282212.1 435 Intron NP_001269141.1
NM_001282213.1 435 Intron NP_001269142.1
NM_001282214.1 435 Intron NP_001269143.1
NM_001282215.1 435 Intron NP_001269144.1
NM_001282216.1 435 Intron NP_001269145.1
NM_001320329.1 435 Intron NP_001307258.1
NM_016250.2 435 Intron NP_057334.1
NM_201535.1 435 Intron NP_963293.1
NM_201536.1 435 Intron NP_963294.1
NM_201537.1 435 Intron NP_963831.1
NM_201538.1 435 Intron NP_963832.1
NM_201539.1 435 Intron NP_963833.1
NM_201540.1 435 Intron NP_963834.1
NM_201541.1 435 Intron NP_963835.1
XM_011536996.2 435 Intron XP_011535298.1
XM_011536997.1 435 Intron XP_011535299.1
XM_011536998.1 435 Intron XP_011535300.1
XM_011536999.1 435 Intron XP_011535301.1
XM_011537001.1 435 Intron XP_011535303.1
XM_011537002.1 435 Intron XP_011535304.1
XM_017021480.1 435 Intron XP_016876969.1
XM_017021481.1 435 Intron XP_016876970.1
XM_017021482.1 435 Intron XP_016876971.1
XM_017021483.1 435 Intron XP_016876972.1
XM_017021484.1 435 Intron XP_016876973.1
XM_017021485.1 435 Intron XP_016876974.1
XM_017021486.1 435 Intron XP_016876975.1
XM_017021487.1 435 Intron XP_016876976.1
XM_017021488.1 435 Intron XP_016876977.1
XM_017021489.1 435 Intron XP_016876978.1
XM_017021490.1 435 Intron XP_016876979.1
XM_017021491.1 435 Intron XP_016876980.1
XM_017021492.1 435 Intron XP_016876981.1
XM_017021493.1 435 Intron XP_016876982.1
XM_017021494.1 435 Intron XP_016876983.1
XM_017021495.1 435 Intron XP_016876984.1
XM_017021496.1 435 Intron XP_016876985.1
XM_017021497.1 435 Intron XP_016876986.1
Gene
RNASE8
Gene Name
ribonuclease A family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138331.2 435 Missense Mutation AAG,GAG K122E NP_612204.1

View Full Product Details