Product Details

SNP ID
rs140860893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100380972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGGCTTCATTGGTAGCGTATGA[C/T]GATTCGGACTCGGAGGCTGAGACAG
Phenotype
MIM: 191050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WARS PubMed Links

Gene Details

Gene
WARS
Gene Name
tryptophanyl-tRNA synthetase
There are no transcripts associated with this gene.

Gene
WDR25
Gene Name
WD repeat domain 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001161476.1 148 Silent Mutation GAC,GAT D16D NP_001154948.1
NM_024515.4 148 Silent Mutation GAC,GAT D16D NP_078791.3
XM_005268056.4 148 Silent Mutation GAC,GAT D16D XP_005268113.1
XM_005268057.3 148 Intron XP_005268114.1
XM_005268058.4 148 Intron XP_005268115.1
XM_011537150.2 148 Silent Mutation GAC,GAT D16D XP_011535452.1
XM_011537151.2 148 Intron XP_011535453.2
XM_017021639.1 148 Intron XP_016877128.1

View Full Product Details