Product Details

SNP ID
rs141081763
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:76769973 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTGTCCCCAGATCCCCATAC[C/T]GAGTGTGCCTACGTTCCAGCCGTCT
Phenotype
MIM: 609011
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
VASH1 PubMed Links

Gene Details

Gene
VASH1
Gene Name
vasohibin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014909.4 282 Missense Mutation CCG,CTG P107L NP_055724.1
XM_017021087.1 282 UTR 5 XP_016876576.1
XM_017021088.1 282 UTR 5 XP_016876577.1
XM_017021089.1 282 UTR 5 XP_016876578.1
XM_017021090.1 282 UTR 5 XP_016876579.1
XM_017021091.1 282 UTR 5 XP_016876580.1

View Full Product Details