Product Details

SNP ID
rs141264462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73478897 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCAAACGTGTCTCATGTGAGC[A/G]GCAGAGCCCAAGTGAAGGCCTCTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HEATR4 PubMed Links

Gene Details

Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 Intron NP_001207413.1
NM_203309.2 Intron NP_976054.2
XM_006720143.2 Intron XP_006720206.1
XM_011536760.2 Intron XP_011535062.1
XM_011536761.2 Intron XP_011535063.1
XM_017021289.1 Intron XP_016876778.1

View Full Product Details