Product Details

SNP ID
rs141786181
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20955958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTCATTAACAATTATCAACGG[C/T]GATGCAAAAACCAAAATACTTTCCT
Phenotype
MIM: 131410
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNASE2 PubMed Links

Gene Details

Gene
RNASE2
Gene Name
ribonuclease A family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002934.2 258 Silent Mutation CGA,TGA R63* NP_002925.1

View Full Product Details