Product Details

SNP ID
rs141843246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60713594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGATAAGAAATTCTCTTTCATG[A/T]TAGCTACCGATTGCAGAACCAAACG
Phenotype
MIM: 606342
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SIX4 PubMed Links

Gene Details

Gene
SIX4
Gene Name
SIX homeobox 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017420.4 2219 Missense Mutation AAC,ATC N720I NP_059116.3
XM_005267759.2 2219 Missense Mutation AAC,ATC N712I XP_005267816.1

View Full Product Details