Product Details

SNP ID
rs141882731
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35087970 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATAACCTTGACATCTTGAAATGAA[A/T]CAGGATCTTGTCCATGGATTTTCAT
Phenotype
MIM: 615902
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM177A1 PubMed Links

Gene Details

Gene
FAM177A1
Gene Name
family with sequence similarity 177 member A1
There are no transcripts associated with this gene.

Gene
LOC101927178
Gene Name
uncharacterized LOC101927178
There are no transcripts associated with this gene.

Gene
PPP2R3C
Gene Name
protein phosphatase 2 regulatory subunit B''gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305155.1 1618 Missense Mutation GAT,GTT D275V NP_001292084.1
NM_001305156.1 1618 Missense Mutation GAT,GTT D275V NP_001292085.1
NM_017917.3 1618 Missense Mutation GAT,GTT D385V NP_060387.2
XM_005267782.3 1618 Missense Mutation GAT,GTT D385V XP_005267839.1
XM_017021388.1 1618 Missense Mutation GAT,GTT D339V XP_016876877.1
XM_017021389.1 1618 Missense Mutation GAT,GTT D333V XP_016876878.1

View Full Product Details