Product Details
- SNP ID
-
rs141963625
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92797845 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATAGTTCACAGAAGGAGCCTACCGG[C/G]AGGGTGGAAATCAGAAAGGAAAAAG
- Phenotype
-
MIM: 606918
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA5
PubMed Links
Gene Details
- Gene
- GOLGA5
- Gene Name
- golgin A5
View Full Product Details