Product Details

SNP ID
rs142171275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57274401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGAGTCGTGATAGCTGTTCACGC[A/G]TCAATAAAACATCCATTTATGGACT
Phenotype
MIM: 614368 MIM: 604469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP5M1 PubMed Links

Gene Details

Gene
AP5M1
Gene Name
adaptor related protein complex 5 mu 1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018229.3 631 Missense Mutation ATC,GTC I78V NP_060699.3
XM_006720196.2 631 UTR 5 XP_006720259.1
XM_011536940.2 631 Missense Mutation ATC,GTC I92V XP_011535242.1
Gene
EXOC5
Gene Name
exocyst complex component 5
There are no transcripts associated with this gene.

View Full Product Details