Product Details

SNP ID
rs142653634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93203400 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAAATCAATGATTATTTTAAAAA[C/T]AGAAAACAATGATAAAAATTCAGTT
Phenotype
MIM: 613816
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GON7 PubMed Links

Gene Details

Gene
GON7
Gene Name
GON7, KEOPS complex subunit homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032490.4 668 UTR 3 NP_115879.2
Gene
UBR7
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
There are no transcripts associated with this gene.

View Full Product Details