Product Details

SNP ID
rs142765501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:63599166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACTACTTCTCTGACATCCTGGAT[A/G]TTCAAACGTGTAACTGGTTCATCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WDR89 PubMed Links

Gene Details

Gene
WDR89
Gene Name
WD repeat domain 89
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008726.2 1539 Silent Mutation AAC,AAT N259N NP_001008726.1
NM_001258272.1 1539 Silent Mutation AAC,AAT N259N NP_001245201.1
NM_080666.3 1539 Silent Mutation AAC,AAT N259N NP_542397.1
XM_011536383.2 1539 Silent Mutation AAC,AAT N297N XP_011534685.1
XM_011536385.2 1539 Silent Mutation AAC,AAT N259N XP_011534687.1

View Full Product Details