Product Details

SNP ID
rs142847094
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20313605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTGATACTGACGATTGCGCTCC[A/T]TAAGTTTCTCAGAGATGTCACTGAA
Phenotype
MIM: 608249
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CCNB1IP1 PubMed Links

Gene Details

Gene
CCNB1IP1
Gene Name
cyclin B1 interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021178.4 983 Missense Mutation AAG,ATG K165M NP_067001.3
NM_182849.2 983 Missense Mutation AAG,ATG K165M NP_878269.1
NM_182852.3 983 Missense Mutation AAG,ATG K165M NP_878272.1
Gene
TTC5
Gene Name
tetratricopeptide repeat domain 5
There are no transcripts associated with this gene.

View Full Product Details