Product Details

SNP ID
rs142972704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:80955648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCTGAGAATGAGGCGATTTCGG[A/G]GGATGGAGAAATAGCCCCGAGTCCC
Phenotype
MIM: 603372
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP128 PubMed Links

Gene Details

Gene
CEP128
Gene Name
centrosomal protein 128
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152446.3 344 Intron NP_689659.2
XM_006720056.3 344 Intron XP_006720119.1
XM_011536490.2 344 Intron XP_011534792.1
XM_011536491.2 344 Intron XP_011534793.1
XM_011536492.2 344 Intron XP_011534794.1
XM_011536493.2 344 Intron XP_011534795.1
XM_011536495.2 344 Intron XP_011534797.1
XM_017021042.1 344 Intron XP_016876531.1
XM_017021043.1 344 Intron XP_016876532.1
Gene
TSHR
Gene Name
thyroid stimulating hormone receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000369.2 344 UTR 5 NP_000360.2
NM_001018036.2 344 UTR 5 NP_001018046.1
NM_001142626.2 344 UTR 5 NP_001136098.1
XM_005268037.4 344 UTR 5 XP_005268094.1
XM_005268039.1 344 UTR 5 XP_005268096.1
XM_006720245.1 344 UTR 5 XP_006720308.1
XM_011537119.2 344 UTR 5 XP_011535421.1

View Full Product Details