Product Details

SNP ID
rs143064811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20801711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCCGGTATGCACAGTTGGGGT[A/G]CCTGGAGCCGTTTGTCAGGCGGCAG
Phenotype
MIM: 180440
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RNASE1 PubMed Links

Gene Details

Gene
RNASE1
Gene Name
ribonuclease A family member 1, pancreatic
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002933.4 535 Missense Mutation CAC,TAC H120Y NP_002924.1
NM_198232.2 535 Missense Mutation CAC,TAC H120Y NP_937875.1
NM_198234.2 535 Missense Mutation CAC,TAC H120Y NP_937877.1
NM_198235.2 535 Missense Mutation CAC,TAC H120Y NP_937878.1

View Full Product Details