Product Details

SNP ID
rs143088473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20368450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGCACATCTTCATTCCCAATTC[A/G]GAAAGTACACATTGCCCTGCACGTC
Phenotype
MIM: 601686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TEP1 PubMed Links

Gene Details

Gene
TEP1
Gene Name
telomerase associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319035.1 6396 Missense Mutation CCG,CTG P2516L NP_001305964.1
NM_007110.4 6396 Missense Mutation CCG,CTG P2624L NP_009041.2
XM_005268027.4 6396 Missense Mutation CCG,CTG P2624L XP_005268084.1
XM_011537110.2 6396 Missense Mutation CCG,CTG P2101L XP_011535412.1

View Full Product Details