Product Details

SNP ID
rs143164089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:39401255 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTCATTTCGGTTGTCACTAAG[A/G]TCTGGAAAACCAGATGTGTCAATCA
Phenotype
MIM: 609103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO33 PubMed Links

Gene Details

Gene
FBXO33
Gene Name
F-box protein 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203301.3 1655 Silent Mutation GAC,GAT D439D NP_976046.1

View Full Product Details