Product Details

SNP ID
rs143769146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:81474340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTCGGCTAACCAACAAACCAAA[C/T]GCACAAGAGAATACTAAACAGGGTT
Phenotype
MIM: 602329
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEL1L PubMed Links

Gene Details

Gene
SEL1L
Gene Name
SEL1L ERAD E3 ligase adaptor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244984.1 5134 Intron NP_001231913.1
NM_005065.5 5134 UTR 3 NP_005056.3
XM_005267988.2 5134 UTR 3 XP_005268045.1
XM_005267989.3 5134 UTR 3 XP_005268046.1

View Full Product Details