Product Details

SNP ID
rs144294831
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74946248 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGTCTCTGCTTCTCTCTCCT[C/G]CTCTTCCCCCTGCCCTTGGGTCTCC
Phenotype
MIM: 601121
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PGF PubMed Links

Gene Details

Gene
PGF
Gene Name
placental growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207012.1 972 Intron NP_001193941.1
NM_001293643.1 972 Missense Mutation AGC,AGG S149R NP_001280572.1
NM_002632.5 972 Missense Mutation AGC,AGG S150R NP_002623.2

View Full Product Details