Product Details

SNP ID
rs144339242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:90401293 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCTTCTCCCTATTTGATAAAGA[C/T]GGCGATGGCACCATCACAACAAAGG
Phenotype
MIM: 114180
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CALM1 PubMed Links

Gene Details

Gene
CALM1
Gene Name
calmodulin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006888.4 946 Silent Mutation GAC,GAT D23D NP_008819.1
XM_006720258.3 946 Silent Mutation GAC,GAT D24D XP_006720321.1

View Full Product Details