Product Details

SNP ID
rs144473753
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95417985 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGTGGGAGCGCCACACAGCA[C/T]GCCCTCCGGAAGAGGGAGCCCAGTC
Phenotype
MIM: 610861
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LINC00341 PubMed Links
Additional Information
For this assay, SNP(s) [rs12434757] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
LINC00341
Gene Name
long intergenic non-protein coding RNA 341
There are no transcripts associated with this gene.

Gene
SYNE3
Gene Name
spectrin repeat containing nuclear envelope family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152592.3 2768 Silent Mutation GCA,GCG A923A NP_689805.3
XM_005267377.3 2768 Silent Mutation GCA,GCG A923A XP_005267434.1
XM_006720063.3 2768 Silent Mutation GCA,GCG A923A XP_006720126.1
XM_011536515.2 2768 Silent Mutation GCA,GCG A931A XP_011534817.1
XM_017021056.1 2768 Silent Mutation GCA,GCG A918A XP_016876545.1
XM_017021057.1 2768 Intron XP_016876546.1

View Full Product Details