Product Details

SNP ID
rs144715100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:103643887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTGCCTGGCCCGAAGGCATTTG[C/T]TTAATTTCCTTTTGCTTCCAATGTG
Phenotype
MIM: 600025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLC1 PubMed Links

Gene Details

Gene
KLC1
Gene Name
kinesin light chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130107.1 Intron NP_001123579.1
NM_005552.4 Intron NP_005543.2
NM_182923.3 Intron NP_891553.2

View Full Product Details