Product Details

SNP ID
rs144807486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23569679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGAATGCCAGGCTGAGGTCCAG[A/G]AGGGCCACGGCTCCCACCACCAGGG
Phenotype
MIM: 603534
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AP1G2 PubMed Links

Gene Details

Gene
AP1G2
Gene Name
adaptor related protein complex 1 gamma 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282474.1 2760 Intron NP_001269403.1
NM_001282475.1 2760 Intron NP_001269404.1
NM_003917.4 2760 Intron NP_003908.1
XM_005268167.3 2760 Intron XP_005268224.1
XM_005268168.4 2760 Intron XP_005268225.1
XM_005268169.3 2760 Intron XP_005268226.1
XM_005268170.3 2760 Intron XP_005268227.1
XM_005268172.3 2760 Intron XP_005268229.1
XM_005268173.3 2760 Intron XP_005268230.1
XM_005268174.3 2760 Intron XP_005268231.1
XM_005268175.3 2760 Intron XP_005268232.1
XM_005268177.3 2760 Intron XP_005268234.1
XM_005268178.3 2760 Intron XP_005268235.1
XM_005268179.3 2760 Intron XP_005268236.1
XM_005268180.4 2760 Intron XP_005268237.1
XM_005268182.3 2760 Intron XP_005268239.1
XM_006720301.3 2760 Intron XP_006720364.1
XM_011537283.2 2760 Intron XP_011535585.1
XM_011537286.2 2760 Intron XP_011535588.1
XM_017021740.1 2760 Intron XP_016877229.1
XM_017021741.1 2760 Intron XP_016877230.1
XM_017021742.1 2760 Intron XP_016877231.1
XM_017021743.1 2760 Intron XP_016877232.1
XM_017021744.1 2760 Intron XP_016877233.1
XM_017021745.1 2760 Intron XP_016877234.1
XM_017021746.1 2760 Intron XP_016877235.1
XM_017021747.1 2760 Intron XP_016877236.1
XM_017021748.1 2760 Intron XP_016877237.1
Gene
JPH4
Gene Name
junctophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146028.1 2760 Silent Mutation CTC,CTT L614L NP_001139500.1
NM_032452.2 2760 Silent Mutation CTC,CTT L614L NP_115828.2
Gene
LOC102724814
Gene Name
uncharacterized LOC102724814
There are no transcripts associated with this gene.

View Full Product Details