Product Details

SNP ID
rs145348711
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93212092 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACAACTTTTTTGGATTGTACTGC[A/T]TTTGCAAGAGACCTTATCCTGATCC
Phenotype
MIM: 613816
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GON7 PubMed Links

Gene Details

Gene
GON7
Gene Name
GON7, KEOPS complex subunit homolog
There are no transcripts associated with this gene.

Gene
UBR7
Gene Name
ubiquitin protein ligase E3 component n-recognin 7 (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_175748.3 642 Missense Mutation ATT,TTT I136F NP_786924.2

View Full Product Details