Product Details

SNP ID
rs145845805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:104987545 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCATGAGAGATCAGAAATAAAAC[C/T]AGTGAGGAAGGAAATAAAGATATTG
Phenotype
MIM: 608570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AHNAK2 PubMed Links

Gene Details

Gene
AHNAK2
Gene Name
AHNAK nucleoprotein 2
There are no transcripts associated with this gene.

Gene
C14orf79
Gene Name
chromosome 14 open reading frame 79
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174891.3 Intron NP_777551.2
XM_005267317.3 Intron XP_005267374.1
XM_005267318.3 Intron XP_005267375.1

View Full Product Details