Product Details
- SNP ID
-
rs145932252
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:94769802 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGCCGAGGCGGGAGCCGCTGACCG[C/T]GGCCGGGAGGCCCGCGCCGCCGGGG
- Phenotype
-
MIM: 138890
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GSC
PubMed Links
Gene Details
- Gene
- GSC
- Gene Name
- goosecoid homeobox
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_173849.2 |
361 |
Missense Mutation |
ACG,GCG |
T72A |
NP_776248.1 |
View Full Product Details