Product Details

SNP ID
rs145971860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50878819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACAGTGAACTTCTACCTGAGA[A/G]TTGAACCTGGGGTGATGCTAGGGAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD12B PubMed Links

Gene Details

Gene
ABHD12B
Gene Name
abhydrolase domain containing 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206673.1 322 Missense Mutation ATT,GTT I103V NP_001193602.1
NM_181533.3 322 UTR 5 NP_853511.2
NM_181814.1 322 Intron NP_861535.1
XM_006720052.1 322 Intron XP_006720115.1
XM_011536472.1 322 Missense Mutation ATT,GTT I103V XP_011534774.1
XM_011536474.2 322 Missense Mutation ATT,GTT I61V XP_011534776.1
XM_017021031.1 322 UTR 5 XP_016876520.1

View Full Product Details