Product Details

SNP ID
rs146076639
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20224194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACTGGGAAGTTCTGTATAATTC[C/T]GGTCTGTGTATGCTGGGTAGGCGGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR11H6 PubMed Links

Gene Details

Gene
OR11H6
Gene Name
olfactory receptor family 11 subfamily H member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004480.1 485 Missense Mutation CCG,CTG P162L NP_001004480.1

View Full Product Details